Product Details

SNP ID
rs535763268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55134225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCTAGGCCCAGGGACGGAGAG[A/G]AACTTGGGCCCAGAGAGGTTGTGGG
Phenotype
MIM: 191041
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNNT1 PubMed Links

Gene Details

Gene
TNNT1
Gene Name
troponin T1, slow skeletal type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126132.2 Intron NP_001119604.1
NM_001126133.2 Intron NP_001119605.1
NM_001291774.1 Intron NP_001278703.1
NM_003283.5 Intron NP_003274.3
XM_011527246.2 Intron XP_011525548.1
XM_017027186.1 Intron XP_016882675.1
XM_017027187.1 Intron XP_016882676.1

View Full Product Details