Product Details

SNP ID
hCV25648181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78624998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCATCCTTGCCTGTTCCACGG[A/C]TGTGGCTGGTTTGATGGGGTTGATG
Phenotype
MIM: 118503 MIM: 118509
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHRNA3 PubMed Links

Gene Details

Gene
CHRNA3
Gene Name
cholinergic receptor nicotinic alpha 3 subunit
There are no transcripts associated with this gene.

Gene
CHRNB4
Gene Name
cholinergic receptor nicotinic beta 4 subunit
There are no transcripts associated with this gene.

View Full Product Details