Product Details

SNP ID
rs10410289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12648277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTCCGCCAGGAGCCGGTGTCC[C/G]GCGGCTGCAGCCTGGGCTGTGTCCA
Phenotype
MIM: 609458
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAN2B1 PubMed Links

Gene Details

Gene
MAN2B1
Gene Name
mannosidase alpha class 2B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000528.3 2217 Silent Mutation GCC,GCG A854A NP_000519.2
NM_001173498.1 2217 Silent Mutation GCC,GCG A853A NP_001166969.1
XM_005259913.1 2217 Silent Mutation GCC,GCG A855A XP_005259970.1
XM_017026818.1 2217 Silent Mutation GCC,GCG A487A XP_016882307.1

View Full Product Details