Product Details

SNP ID
rs55823456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57506390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGGAGCCAAGGCTGAGGCAGCT[C/G]CTGAGCAGAGTGCTTCTGTAGAAGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF773 PubMed Links

Gene Details

Gene
ZNF773
Gene Name
zinc finger protein 773
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304334.1 519 Missense Mutation CCT,GCT P98A NP_001291263.1
NM_001304335.1 519 Intron NP_001291264.1
NM_001304336.1 519 Silent Mutation CTC,CTG L96L NP_001291265.1
NM_001304337.1 519 Intron NP_001291266.1
NM_198542.2 519 Missense Mutation CCT,GCT P99A NP_940944.1

View Full Product Details