Product Details

SNP ID
rs35809233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139266925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGACAGGCAGATGCCTCTTTTA[C/T]GACCACAACTGTAGCACTGGGACTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGARP PubMed Links

Gene Details

Gene
MGARP
Gene Name
mitochondria localized glutamic acid rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032623.3 577 Missense Mutation ATA,GTA I133V NP_116012.2

View Full Product Details