Product Details
- SNP ID
-
rs567486188
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:32918284 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTTATATAAGTTAAAACTTTAAA[C/T]AGATACTATCACCAGGACAACCTCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RP9P
PubMed Links
Gene Details
- Gene
- RP9P
- Gene Name
- retinitis pigmentosa 9 pseudogene
There are no transcripts associated with this gene.
View Full Product Details