Product Details

SNP ID
rs17854363
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:143356630 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACGTCAGACACCTTCCGGCTGAGAG[C/T]GGTAGACATCTCAGGGTCTTCTCGT
Phenotype
MIM: 118425
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCN1 PubMed Links

Gene Details

Gene
CLCN1
Gene Name
chloride voltage-gated channel 1
There are no transcripts associated with this gene.

Gene
FAM131B
Gene Name
family with sequence similarity 131 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031690.2 1139 Missense Mutation ACT,GCT T335A NP_001026860.2
NM_001278297.1 1139 Missense Mutation ACT,GCT T241A NP_001265226.1
NM_014690.4 1139 Missense Mutation ACT,GCT T307A NP_055505.3
XM_005250073.3 1139 Missense Mutation ACT,GCT T307A XP_005250130.1
XM_006716186.2 1139 Missense Mutation ACT,GCT T275A XP_006716249.1

View Full Product Details