Product Details

SNP ID
rs61730145
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219181766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACAACTCCGCAGCTGACTGAT[A/G]TCTCCGAGGGTATGGGGAGCCCTTT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CNPPD1 PubMed Links

Gene Details

Gene
CNPPD1
Gene Name
cyclin Pas1/PHO80 domain containing 1
There are no transcripts associated with this gene.

Gene
FAM134A
Gene Name
family with sequence similarity 134 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321109.1 1169 Missense Mutation ATC,GTC I129V NP_001308038.1
NM_001321110.1 1169 Missense Mutation ATC,GTC I129V NP_001308039.1
NM_024293.5 1169 Missense Mutation ATC,GTC I336V NP_077269.3
XM_005246848.2 1169 Missense Mutation ATC,GTC I336V XP_005246905.1

View Full Product Details