Product Details

SNP ID
rs61733724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74874248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCAGGCAGTGGCACCCACACCC[A/G]GTGGGACACACCTCCTGGGTCCGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR59 PubMed Links

Gene Details

Gene
WDR59
Gene Name
WD repeat domain 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324171.1 2730 Intron NP_001311100.1
NM_001324172.1 2730 Intron NP_001311101.1
NM_030581.3 2730 Silent Mutation ACC,ACT T962T NP_085058.3
XM_005256146.3 2730 Silent Mutation ACC,ACT T981T XP_005256203.1
XM_011523331.2 2730 Silent Mutation ACC,ACT T674T XP_011521633.1
XM_011523332.2 2730 Silent Mutation ACC,ACT T660T XP_011521634.1
XM_017023667.1 2730 Silent Mutation ACC,ACT T710T XP_016879156.1
XM_017023668.1 2730 Silent Mutation ACC,ACT T710T XP_016879157.1
XM_017023669.1 2730 Silent Mutation ACC,ACT T674T XP_016879158.1
XM_017023670.1 2730 Silent Mutation ACC,ACT T660T XP_016879159.1

View Full Product Details