Product Details

SNP ID
rs61733725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74874253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGTGGCACCCACACCCGGTGG[A/G]ACACACCTCCTGGGTCCGAAACCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR59 PubMed Links

Gene Details

Gene
WDR59
Gene Name
WD repeat domain 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324171.1 2725 Intron NP_001311100.1
NM_001324172.1 2725 Intron NP_001311101.1
NM_030581.3 2725 Missense Mutation CCC,TCC P961S NP_085058.3
XM_005256146.3 2725 Missense Mutation CCC,TCC P980S XP_005256203.1
XM_011523331.2 2725 Missense Mutation CCC,TCC P673S XP_011521633.1
XM_011523332.2 2725 Missense Mutation CCC,TCC P659S XP_011521634.1
XM_017023667.1 2725 Missense Mutation CCC,TCC P709S XP_016879156.1
XM_017023668.1 2725 Missense Mutation CCC,TCC P709S XP_016879157.1
XM_017023669.1 2725 Missense Mutation CCC,TCC P673S XP_016879158.1
XM_017023670.1 2725 Missense Mutation CCC,TCC P659S XP_016879159.1

View Full Product Details