Product Details

SNP ID
rs61735331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6621879 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACCCCAAATGGTGAGACAACG[A/G]GTGAGCGAGCAGCCAGAGGAGACAG
Phenotype
MIM: 603057 MIM: 600475 MIM: 607998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCHS1 PubMed Links

Gene Details

Gene
DCHS1
Gene Name
dachsous cadherin-related 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003737.3 10204 Missense Mutation CCC,CTC P3266L NP_003728.1
Gene
TAF10
Gene Name
TATA-box binding protein associated factor 10
There are no transcripts associated with this gene.

Gene
TPP1
Gene Name
tripeptidyl peptidase 1
There are no transcripts associated with this gene.

View Full Product Details