Product Details

SNP ID
rs34958581
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:32471206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGTGCCTGAGAGAGTCCCCTTG[C/T]GGTGACTGCGGAGACTGCCACCCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB8B PubMed Links

Gene Details

Gene
ZBTB8B
Gene Name
zinc finger and BTB domain containing 8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145720.1 672 Silent Mutation TGC,TGT C194C NP_001139192.1

View Full Product Details