Product Details

SNP ID
rs10415207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12351257 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTACATGGATCTACTCCAGGAG[G/T]TTTTTCATTCACAGTACTATCTGGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF442 PubMed Links

Gene Details

Gene
ZNF442
Gene Name
zinc finger protein 442
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030824.2 367 Missense Mutation ACT,CCT T110P NP_110451.1
XM_006722908.3 367 Missense Mutation ACT,CCT T103P XP_006722971.1
XM_017027316.1 367 Missense Mutation ACT,CCT T118P XP_016882805.1
XM_017027317.1 367 Missense Mutation ACT,CCT T53P XP_016882806.1
XM_017027318.1 367 Missense Mutation ACT,CCT T110P XP_016882807.1
Gene
ZNF563
Gene Name
zinc finger protein 563
There are no transcripts associated with this gene.

View Full Product Details