Product Details

SNP ID
rs35273540
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33292438 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTCATGGCCTCTGACCTCGCTCC[C/T]CTGGTAGCAGCTGTACCAGCTCATA
Phenotype
MIM: 605660 MIM: 602306 MIM: 601962 MIM: 611440
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MIR6834 PubMed Links
Additional Information
For this assay, SNP(s) [rs149708654] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MIR6834
Gene Name
microRNA 6834
There are no transcripts associated with this gene.

Gene
MIR6873
Gene Name
microRNA 6873
There are no transcripts associated with this gene.

Gene
PFDN6
Gene Name
prefoldin subunit 6
There are no transcripts associated with this gene.

Gene
RGL2
Gene Name
ral guanine nucleotide dissociation stimulator like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243738.1 2477 Missense Mutation GAG,GGG E623G NP_001230667.1
NM_004761.4 2477 Missense Mutation GAG,GGG E705G NP_004752.1
XM_011514781.1 2477 Missense Mutation GAG,GGG E598G XP_011513083.1
XM_011514785.2 2477 Intron XP_011513087.1
Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
There are no transcripts associated with this gene.

Gene
WDR46
Gene Name
WD repeat domain 46
There are no transcripts associated with this gene.

View Full Product Details