Product Details

SNP ID
rs7079652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14966555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAAATCTGTACAGATTTCAACA[A/G]GCAGGATATCGGGATGAAACCGAAT
Phenotype
MIM: 614174
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105376430 PubMed Links

Gene Details

Gene
LOC105376430
Gene Name
uncharacterized LOC105376430
There are no transcripts associated with this gene.

Gene
MEIG1
Gene Name
meiosis/spermiogenesis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080836.2 235 Silent Mutation CAA,CAG Q29Q NP_001074305.1

View Full Product Details