Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001185181.2 | Intron | NP_001172110.1 | |||
NM_001265595.1 | Intron | NP_001252524.1 | |||
NM_001265596.1 | Intron | NP_001252525.1 | |||
NM_014260.3 | Intron | NP_055075.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001164267.1 | Intron | NP_001157739.1 | |||
NM_005452.5 | Intron | NP_005443.3 | |||
XM_017011484.1 | Intron | XP_016866973.1 | |||
XM_017011485.1 | Intron | XP_016866974.1 | |||
XM_017011486.1 | Intron | XP_016866975.1 | |||
XM_017011487.1 | Intron | XP_016866976.1 |