Product Details

SNP ID
rs7258170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57193913 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGATGATGTGGCTGTGACTTTCAC[C/T]AAGGAGGAGTGGGGGCAGCTGGACC
Phenotype
MIM: 604668
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF264 PubMed Links

Gene Details

Gene
ZNF264
Gene Name
zinc finger protein 264
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003417.4 486 Silent Mutation ACC,ACT T24T NP_003408.1
XM_011527522.2 486 Silent Mutation ACC,ACT T24T XP_011525824.1

View Full Product Details