Product Details

SNP ID
hCV25652946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2818000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGCGAACGGAGGGGTGCGGGGA[A/C]GGGCAGGAACAGGGCTGGAGGGAGT
Phenotype
MIM: 608159
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRSS21 PubMed Links

Gene Details

Gene
PRSS21
Gene Name
protease, serine 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270452.1 Intron NP_001257381.1
NM_006799.3 Intron NP_006790.1
NM_144956.2 Intron NP_659205.1
NM_144957.2 Intron NP_659206.1

View Full Product Details