Product Details

SNP ID
rs191985514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123417010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCAACCGGGCCCCGCCTCTTCCC[G/T]GCCCCTCCTCGGGGGCGCTCCCGGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WDYHV1 PubMed Links

Gene Details

Gene
WDYHV1
Gene Name
WDYHV motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283024.1 Intron NP_001269953.1
NM_001283027.1 Intron NP_001269956.1
NM_018024.2 Intron NP_060494.1
XM_006716597.3 Intron XP_006716660.1
XM_011517147.1 Intron XP_011515449.1
XM_011517151.2 Intron XP_011515453.1
XM_017013601.1 Intron XP_016869090.1
XM_017013602.1 Intron XP_016869091.1
XM_017013603.1 Intron XP_016869092.1

View Full Product Details