Product Details

SNP ID
rs61736384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1434624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAGGGGCTGCTTTTGCTTTTT[A/G]GTGACCAGATCTTGGACAAGCGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MICALL2 PubMed Links

Gene Details

Gene
MICALL2
Gene Name
MICAL like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182924.3 2811 Missense Mutation CCA,CTA P896L NP_891554.1
XM_011515520.2 2811 Silent Mutation ACC,ACT T894T XP_011513822.1
XM_011515522.2 2811 Silent Mutation ACC,ACT T880T XP_011513824.1
XM_011515523.2 2811 Silent Mutation ACC,ACT T811T XP_011513825.1
XM_011515524.2 2811 Silent Mutation ACC,ACT T323T XP_011513826.1
XM_017012619.1 2811 Intron XP_016868108.1

View Full Product Details