Product Details
- SNP ID
-
rs3803869
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44559686 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTGTGTGTGTTGGGGAGTGTAGTTG[G/T]GGGGAGGGTTCTCTTTCTTAAGTGC
- Phenotype
-
MIM: 600667
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FZD2
PubMed Links
Gene Details
- Gene
- FZD2
- Gene Name
- frizzled class receptor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001466.3 |
2243 |
UTR 3 |
|
|
NP_001457.1 |
View Full Product Details