Product Details
- SNP ID
-
rs113862234
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:868031 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GCTTGTCCAGGTTCAGCGCTCTCCG[C/T]GGGTGAGGCAAGGAAACCGAGGAGA
- Phenotype
-
MIM: 134350
MIM: 604062
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CFD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117801197] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CFD
- Gene Name
- complement factor D
There are no transcripts associated with this gene.
- Gene
- MED16
- Gene Name
- mediator complex subunit 16
View Full Product Details