Product Details

SNP ID
rs142247598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:67526367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAGTGTTATACGTGATCAGTAA[C/T]TTGCATGATTAATAAGCATTGATGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C15orf61 PubMed Links

Gene Details

Gene
C15orf61
Gene Name
chromosome 15 open reading frame 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143936.1 Intron NP_001137408.1
Gene
IQCH-AS1
Gene Name
IQCH antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details