Product Details

SNP ID
hCV25742379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:1083054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGCAGGGACAGTGCAGGCCTCA[G/C]CCCTCACCGGAGGCTCTCGGGAGTG
Phenotype
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf159 PubMed Links

Gene Details

Gene
C1orf159
Gene Name
chromosome 1 open reading frame 159
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017891.4 Intron NP_060361.4
Gene
RNF223
Gene Name
ring finger protein 223
There are no transcripts associated with this gene.

View Full Product Details