Product Details

SNP ID
rs111287000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184249681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGGCCGGCGGCTCTGGCTGCC[A/C]GGCGGTTGAGAGCATGGCCTCTCCA
Phenotype
MIM: 608750 MIM: 608721 MIM: 610145 MIM: 611620
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALG3 PubMed Links

Gene Details

Gene
ALG3
Gene Name
ALG3, alpha-1,3- mannosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006941.2 25 Intron NP_001006942.1
NM_005787.5 25 Intron NP_005778.1
XM_011512322.1 25 Intron XP_011510624.1
XM_011512323.2 25 Intron XP_011510625.1
Gene
CAMK2N2
Gene Name
calcium/calmodulin dependent protein kinase II inhibitor 2
There are no transcripts associated with this gene.

Gene
ECE2
Gene Name
endothelin converting enzyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037324.2 25 Intron NP_001032401.1
NM_001100120.1 25 Intron NP_001093590.1
NM_001100121.1 25 Intron NP_001093591.1
NM_014693.3 25 UTR 5 NP_055508.3
NM_032331.3 25 UTR 5 NP_115707.2
Gene
MIR1224
Gene Name
microRNA 1224
There are no transcripts associated with this gene.

Gene
VWA5B2
Gene Name
von Willebrand factor A domain containing 5B2
There are no transcripts associated with this gene.

View Full Product Details