Product Details

SNP ID
rs61745347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:8871251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTGAGGCCGCCTCCATAGCCAC[C/T]GTGCGCCCGTGGGTACCAGCGCGCT
Phenotype
MIM: 142992
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMX1 PubMed Links

Gene Details

Gene
HMX1
Gene Name
H6 family homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306142.1 567 Missense Mutation AGT,GGT S122G NP_001293071.1
NM_018942.2 567 Missense Mutation AGT,GGT S122G NP_061815.2

View Full Product Details