Product Details

SNP ID
rs4795938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34581045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCAAGTCGTCGTCGACTGTTGC[C/T]CTCTCGGACCCCTCCCGCCCCCGCC
Phenotype
MIM: 616178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf102 PubMed Links

Gene Details

Gene
C17orf102
Gene Name
chromosome 17 open reading frame 102
There are no transcripts associated with this gene.

Gene
TMEM132E
Gene Name
transmembrane protein 132E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304438.1 297 UTR 5 NP_001291367.1

View Full Product Details