Product Details

SNP ID
rs61744609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151198917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTGCCCGGAACTCCTCCTCTTT[A/C]TCCATCTGGCGGAGCCTCTGGACAT
Phenotype
MIM: 612510
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABCF2 PubMed Links

Gene Details

Gene
ABCF2
Gene Name
ATP binding cassette subfamily F member 2
There are no transcripts associated with this gene.

Gene
IQCA1L
Gene Name
IQ motif containing with AAA domain 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304419.1 846 Missense Mutation GAG,GAT E268D NP_001291348.1

View Full Product Details