Product Details

SNP ID
rs115862299
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:1669706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGACGAGCATGGCCACCGCAGC[G/A]GCGCTGGTGTAGAACTGCAGCTCCG
Phenotype
MIM: 176873
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CDK11B PubMed Links

Gene Details

Gene
CDK11B
Gene Name
cyclin dependent kinase 11B
There are no transcripts associated with this gene.

Gene
SLC35E2B
Gene Name
solute carrier family 35 member E2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110781.2 1713 Silent Mutation GCC,GCT A264A NP_001104251.1
NM_001290264.1 1713 Silent Mutation GCC,GCT A264A NP_001277193.1
XM_011542072.1 1713 Intron XP_011540374.1

View Full Product Details