Product Details

SNP ID
rs151307272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:176328128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAATAAAGTATTTGACATCTCAG[C/T]GTTTGTCTTAAAATTTTTTTTGAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPCS3 PubMed Links

Gene Details

Gene
SPCS3
Gene Name
signal peptidase complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021928.3 Intron NP_068747.1

View Full Product Details