Product Details

SNP ID
rs61742762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56641563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTTTTAAACTATAGATGAGGG[G/T]ATTTAGCACAGGGATTATTACTGTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR5AP2 PubMed Links

Gene Details

Gene
OR5AP2
Gene Name
olfactory receptor family 5 subfamily AP member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002925.1 877 Missense Mutation ACC,CCC T293P NP_001002925.1

View Full Product Details