Product Details

SNP ID
rs34970007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72139687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCCTCTGGAGCCACTCCTTCAA[A/G]GTCAGCAACTATAGTCGAGGGAGCG
Phenotype
MIM: 602469
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOLR3 PubMed Links

Gene Details

Gene
FOLR3
Gene Name
folate receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000804.3 787 Silent Mutation AAA,AAG K198K NP_000795.2
NM_001318045.1 787 Silent Mutation AAA,AAG K107K NP_001304974.1
XM_011544876.1 787 Silent Mutation AAA,AAG K106K XP_011543178.1

View Full Product Details