Product Details

SNP ID
rs61746300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135051363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGATGGCGATGGCAGTGGTAGC[A/G]TAGGTCTGTGGGCAGAATGATGTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM127B PubMed Links

Gene Details

Gene
FAM127B
Gene Name
family with sequence similarity 127 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001078172.1 281 UTR 3 NP_001071640.1
NM_001134321.1 281 Silent Mutation TAC,TAT Y66Y NP_001127793.1

View Full Product Details