Product Details

SNP ID
rs113498250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57320851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCGCTAGGGCTGCAGGGTCTC[A/G]GGATGGCAGCCTCGGCGCAGGTGAG
Phenotype
MIM: 616847
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF543 PubMed Links

Gene Details

Gene
ZNF543
Gene Name
zinc finger protein 543
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213598.3 355 UTR 5 NP_998763.2

View Full Product Details