Product Details

SNP ID
rs61742526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179837694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAAGAGACCTTGGCTGCTCACT[A/G]TCCACATGTGAACTTTTTCTAGGTG
Phenotype
MIM: 601530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C5orf45 PubMed Links

Gene Details

Gene
C5orf45
Gene Name
chromosome 5 open reading frame 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017987.2 608 Silent Mutation GAC,GAT D188D NP_001017987.1
NM_016175.3 608 Silent Mutation GAC,GAT D243D NP_057259.2
Gene
SQSTM1
Gene Name
sequestosome 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142298.1 608 UTR 3 NP_001135770.1
NM_001142299.1 608 UTR 3 NP_001135771.1
NM_003900.4 608 UTR 3 NP_003891.1
XM_011534683.2 608 Intron XP_011532985.1
XM_017010010.1 608 Intron XP_016865499.1

View Full Product Details