Product Details

SNP ID
rs35060029
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:160031158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGTCATTGTCGCGATCTGGAAG[A/C]AGGTGGAGGGTTATGGGAAGGATGT
Phenotype
MIM: 602208 MIM: 610273
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNJ10 PubMed Links

Gene Details

Gene
KCNJ10
Gene Name
potassium voltage-gated channel subfamily J member 10
There are no transcripts associated with this gene.

Gene
PIGM
Gene Name
phosphatidylinositol glycan anchor biosynthesis class M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145167.2 836 Silent Mutation CTG,CTT L194L NP_660150.1

View Full Product Details