Product Details
- SNP ID
-
rs112115396
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
17
- Location
-
Chr.1:203771380 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGCCTTGCAGGACAAGTTCCTTA[C/T]CTCCGAGTTACCGTCATGCCCTTGC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LAX1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78993747] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LAX1
- Gene Name
- lymphocyte transmembrane adaptor 1
View Full Product Details