Product Details

SNP ID
rs188778961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17553692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGAGCTGGGGACTCCACAGCT[A/C]TCGCTCACCCACTCACCCTGGCCAT
Phenotype
MIM: 604487 MIM: 605242
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OTOG PubMed Links

Gene Details

Gene
OTOG
Gene Name
otogelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277269.1 Intron NP_001264198.1
NM_001292063.1 Intron NP_001278992.1
Gene
USH1C
Gene Name
USH1 protein network component harmonin
There are no transcripts associated with this gene.

View Full Product Details