Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001278253.1 | Intron | NP_001265182.1 | |||
NM_003558.3 | Intron | NP_003549.1 | |||
XM_005252261.3 | Intron | XP_005252318.1 | |||
XM_005252262.4 | Intron | XP_005252319.1 | |||
XM_006717300.2 | Intron | XP_006717363.1 | |||
XM_006717301.1 | Intron | XP_006717364.1 | |||
XM_011519082.2 | Intron | XP_011517384.1 | |||
XM_011519083.2 | Intron | XP_011517385.1 | |||
XM_011519084.2 | Intron | XP_011517386.1 | |||
XM_017015188.1 | Intron | XP_016870677.1 | |||
XM_017015189.1 | Intron | XP_016870678.1 | |||
XM_017015190.1 | Intron | XP_016870679.1 | |||
XM_017015191.1 | Intron | XP_016870680.1 |