Product Details

SNP ID
rs61995677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:24808710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAAAGAGCTTGCATTGGTTCCC[A/G]CGGCTCTGATTTGCTGGGGAGTTGA
Phenotype
MIM: 185490
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC149 PubMed Links

Gene Details

Gene
CCDC149
Gene Name
coiled-coil domain containing 149
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005248210.3 1385 Silent Mutation CGC,CGT R434R XP_005248267.1
XM_011513906.2 1385 Silent Mutation CGC,CGT R434R XP_011512208.1
XM_011513907.2 1385 Silent Mutation CGC,CGT R379R XP_011512209.1
XM_011513908.1 1385 Silent Mutation CGC,CGT R379R XP_011512210.1
XM_017008827.1 1385 Silent Mutation CGC,CGT R423R XP_016864316.1
XM_017008828.1 1385 Silent Mutation CGC,CGT R368R XP_016864317.1
Gene
SOD3
Gene Name
superoxide dismutase 3, extracellular
There are no transcripts associated with this gene.

View Full Product Details