Product Details

SNP ID
rs116600375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2830628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGAGGGGTGGGGTCCCCTGTTC[C/T]GGCAGAGGTCACCCCCATATCACCG
Phenotype
MIM: 608159
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS21 PubMed Links

Gene Details

Gene
PRSS21
Gene Name
protease, serine 21
There are no transcripts associated with this gene.

Gene
PRSS30P
Gene Name
protease, serine, 30 pseudogene
There are no transcripts associated with this gene.

Gene
ZG16B
Gene Name
zymogen granule protein 16B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145252.2 Intron NP_660295.2

View Full Product Details