Product Details

SNP ID
rs73403243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99479538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAGGAGCCCTGAATATATTCCT[C/T]ATGAATTGGGTGCCCTTGGCCCACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF394 PubMed Links

Gene Details

Gene
ZNF394
Gene Name
zinc finger protein 394
There are no transcripts associated with this gene.

Gene
ZNF789
Gene Name
zinc finger protein 789
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013258.1 Intron NP_001013276.1
NM_213603.2 Intron NP_998768.2
XM_011516067.2 Intron XP_011514369.1
XM_011516070.2 Intron XP_011514372.1
XM_011516073.2 Intron XP_011514375.1
XM_017012018.1 Intron XP_016867507.1
XM_017012019.1 Intron XP_016867508.1
XM_017012020.1 Intron XP_016867509.1
XM_017012021.1 Intron XP_016867510.1
XM_017012022.1 Intron XP_016867511.1
XM_017012023.1 Intron XP_016867512.1
XM_017012024.1 Intron XP_016867513.1
XM_017012025.1 Intron XP_016867514.1
XM_017012026.1 Intron XP_016867515.1
XM_017012027.1 Intron XP_016867516.1

View Full Product Details