Product Details

SNP ID
rs61998167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74498086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCGAGGCGACGTCGGCGCGCAT[C/G]TCCTCCACATCGCGCTTGAGCTTGG
Phenotype
MIM: 607164 MIM: 610231
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LBX2 PubMed Links

Gene Details

Gene
LBX2
Gene Name
ladybird homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009812.1 883 Missense Mutation GAC,GAG D142E NP_001009812.1
NM_001282430.1 883 Missense Mutation GAC,GAG D146E NP_001269359.1
Gene
LBX2-AS1
Gene Name
LBX2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PCGF1
Gene Name
polycomb group ring finger 1
There are no transcripts associated with this gene.

Gene
TTC31
Gene Name
tetratricopeptide repeat domain 31
There are no transcripts associated with this gene.

View Full Product Details