Product Details
- SNP ID
-
rs115065558
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:44624569 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAAACCTTGGCTATGATTGTGCA[C/T]GTGGGGTCATTGAGAGCACACAGAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
YIPF7
PubMed Links
Gene Details
- Gene
- YIPF7
- Gene Name
- Yip1 domain family member 7
View Full Product Details