Product Details

SNP ID
rs35075949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27273134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAGACTATGACAGCTACTTGTC[C/T]ATCCGACCCCTTTATCAGCACATCC
Phenotype
MIM: 612403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RASL11A PubMed Links

Gene Details

Gene
RASL11A
Gene Name
RAS like family 11 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206827.1 987 Silent Mutation TCC,TCT S123S NP_996563.1

View Full Product Details