Product Details

SNP ID
rs34927843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:50072705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCCTACCAATGTTGACCTGGCT[A/G]TTTCAGTAGGTAAGTGAAGAAAAGG
Phenotype
MIM: 614951
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEATR3 PubMed Links

Gene Details

Gene
HEATR3
Gene Name
HEAT repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005256013.2 665 Missense Mutation ATT,GTT I119V XP_005256070.1
XM_011523188.2 665 Missense Mutation ATT,GTT I88V XP_011521490.1
XM_017023385.1 665 Intron XP_016878874.1
XM_017023386.1 665 Intron XP_016878875.1

View Full Product Details