Product Details

SNP ID
rs115066153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20292239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCAAAATACTGGACTTAAAGTCA[C/G]TGACAGGTAATTAAAATAAAATATA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TTC5 PubMed Links

Gene Details

Gene
TTC5
Gene Name
tetratricopeptide repeat domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138376.2 Intron NP_612385.2

View Full Product Details