Product Details

SNP ID
rs115401381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66909943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCCCTCCCCTTGCATCCCAGC[A/G]GTTCCCTCAGGAACTGCAGGCTGCA
Phenotype
MIM: 603118
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH16 PubMed Links

Gene Details

Gene
CDH16
Gene Name
cadherin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204744.1 Intron NP_001191673.1
NM_001204745.1 Intron NP_001191674.1
NM_001204746.1 Intron NP_001191675.1
NM_004062.3 Intron NP_004053.1
XM_005255770.2 Intron XP_005255827.1
XM_011522807.1 Intron XP_011521109.1

View Full Product Details