Product Details

SNP ID
rs747191783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68896504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCCTGAATATCCAGAGAAGCTC[A/G]GTGGTCTCACCTTCTCCAGTCGAAT
Phenotype
MIM: 600502 MIM: 611834
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGHMBP2 PubMed Links

Gene Details

Gene
IGHMBP2
Gene Name
immunoglobulin mu binding protein 2
There are no transcripts associated with this gene.

Gene
MRPL21
Gene Name
mitochondrial ribosomal protein L21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181514.1 Intron NP_852615.1
NM_181515.1 Intron NP_852616.1
XM_005273823.4 Intron XP_005273880.1

View Full Product Details