Product Details

SNP ID
rs35195217
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49636368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGGTTGACAGTTATGTTTTTAT[C/A]CAAGGCGTCGTATGTGATGTTTGCA
Phenotype
MIM: 604016
Polymorphism
C/A, Transversion Substitution
Allele Nomenclature
Literature Links
B4GALT5 PubMed Links

Gene Details

Gene
B4GALT5
Gene Name
beta-1,4-galactosyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004776.3 1305 Missense Mutation GAT,TAT D371Y NP_004767.1

View Full Product Details